Linearity LQ hs-CRP for Ortho Vitros, AUDIT MircoControls
76558-072
:
- Description:Linearity LQ hs-CRP for ortho vitros
- Size:1 ml
- Cat. No.:76558-072
The Linearity LQ hsCRP for Ortho Vitros is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for hsCRP.