Order Entry
United States
ContactUsLinkComponent
L-Photo-Leucine, Pierce™
L-Photo-Leucine, Pierce™
Catalog # PI22610
Supplier:  Invitrogen
CAS Number:  
undefined
L-Photo-Leucine, Pierce™
Catalog # PI22610
Supplier:  Invitrogen
Supplier Number:  22610
CAS Number:  

Specifications

  • Size:
    100 mg
  • Cat. No.:
    PI22610
  • Supplier no.:
    22610

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling, Metabolic Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Diazirine
  • Spacer:
    Short
  • Form:
    Solid
  • Quantity:
    100 mg
  • Type:
    Crosslinker
  • Solubility:
    Water

Specification Test Results

About this item

 :
 :143.17 g/mol
Storage Temperature:Refrigerator
Formula:C₅H₉O₂N₃