Flex Bench MS for HPLC, Agilent Technologies
Catalog # AGG6015B
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Specifications
- Description:InfinityLab Flex Bench MS, with power strip
- Cat. No.:AGG6015B
Specifications
About this item
Optimize placement of and access to InfinityLab LC/MSD Series and Ultivo LC/TQ systems.
- Optimize your whole lab setup
- Keep your Agilent InfinityLab MSDs and Ultivo LC/MS systems in one stack, with easy access to all areas
- Improve the lab environment with an integrated solution for waste management and a quiet cover to reduce pump noise
- Engineered to provide an adjustable and safe way to optimize space and to configure your LC system to suit your needs for higher productivity
- Special attention on product ergonomics for a better lab productivity with a safe and healthy environment
- Safely configure your instrument to suit the way you work
The InfinityLab Flex Bench family provides easy ways to configure your instrument to suit your needs, anywhere in the lab, and address the high demand on flexibility in the lab. The Flex Bench MS allows you to optimize your whole lab setup and keep your InfinityLab LC/MSD Series and Ultivo LC/TQ systems in one stack with easy access to all areas.