Hypodermic Needle Set, Mortech
Catalog # 76549-786
Supplier: Avantor
Specifications
- Description:Hypodermic needle set, 10 pc.
- Cat. No.:76549-786
Specifications
About this item
This hypodermic needle set is designed for aspiration/cavity injection or extraction.
Delivery: (1) 1.5" 20 gauge hypodermic needle; (1) 2.5" 20 gauge hypodermic needle; (1) 3" 19 gauge hypodermic needle; (1) 3.5" 15 gauge hypodermic needle; (1) 4" 15 gauge hypodermic needle; (1) 5" 15 gauge hypodermic needle; (1) 6" 12 gauge hypodermic needle; (1) 6" 15 gauge hypodermic needle; (1) 8" 13 gauge hypodermic needle.