ContactUsLinkComponent
Mopec Safe Seal System, Pre-Cut Bags Starter Pack, Mortech
Mopec Safe Seal System, Pre-Cut Bags Starter Pack, Mortech
Catalog # 76549-648
Supplier:  Mopec
undefined
Mopec Safe Seal System, Pre-Cut Bags Starter Pack, Mortech
Catalog # 76549-648
Supplier:  Mopec
Supplier Number:  BE160
Specifications
  • Dimensions:
    48×96"
  • Cat. No.:
    76549-648
Specifications
About this item
Mopec Safe Seal System is a military tested, heavy duty, hermetically sealable Bio-Safety Level 4 (BSL-4) Containment body bag system.
It is easy to use, as the body is placed in the flexible material, then heat-sealed with the Safe Seal Heat Sealer to provide complete and secure containment of infectious human or animal remains. The Safe Seal System was designed to help keep workers and survivors safe after a mass fatality event or any high-risk biohazard exposure.
Delivery: The Starter Pack contains 25 pre-cut 48×96" bags (BE161), 1 heat sealer (BE153), and 2 alignment clips (BE900).