ContactUsLinkComponent
Scalpel Handle, Post Mortem, PM40H, Metal, Mortech
Scalpel Handle, Post Mortem, PM40H, Metal, Mortech
Catalog # 76549-428
Supplier:  Mopec
undefined
Scalpel Handle, Post Mortem, PM40H, Metal, Mortech
Catalog # 76549-428
Supplier:  Mopec
Supplier Number:  AJ125
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Scalpel handle, post mortem, PM40H, metal
  • Cat. No.:
    76549-428
Specifications
About this item
PM40 handle with its heavy duty screw fitting can be used with either a PM40 or PM40 Bull Nose pathology blade.
  • Metal
  • Compatible with round tip (AJ124) blades