ContactUsLinkComponent
Knife Sharpener, 3 Step, Mortech
Knife Sharpener, 3 Step, Mortech
Catalog # 76549-328
undefined
Knife Sharpener, 3 Step, Mortech
Catalog # 76549-328
Supplier Number:  AH030
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Knife Sharpener, 3 Step
  • Length:
    12.5"
  • Cat. No.:
    76549-328
Specifications
About this item
Mopec’s three-step knife sharpener is constructed of durable polypropylene.
  • Includes three changeable ceramic rods and complete instructions
Sharpen virtually any dull knife with the ceramic rods standing vertically or laying horizontally on your bench top. The AH030 can be used for dull or worn edges, partial dull edges and quick touch ups.
Requires no oil or water. Rods can be rotated for sharp new surfaces providing longer stone life.