ContactUsLinkComponent
Forceps, ProCUT Kit, Mortech
Forceps, ProCUT Kit, Mortech
Catalog # 76548-766
undefined
Forceps, ProCUT Kit, Mortech
Catalog # 76548-766
Supplier Number:  AB079
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    33×9 cm (13×3.5" )
  • Cat. No.:
    76548-766
Specifications
About this item
ProCUT is an innovative tool that assists in cutting in parallel slices of uniform thickness.
  • Laser shaped AISI 304 stainless steel
  • 5 mm thickness slice
  • PTFE positioning base
ProCUT eliminates the guesswork in cutting a specific thickness of tissue mass. Suitable for use with both conventional and microwave tissue processors. Serial wet tissue slices are taken in a simple step with the unique design of ProCUT. Movable forceps prongs designed for ergonomic handling of varying sized tissues. Metal side plates secure sample in position. No more complaints about slicing being “too thick” or “not parallel”.