Orders LinkContactUsLinkComponent
Forceps, Dressing, Mortech
Forceps, Dressing, Mortech
6"
Catalog # 76548-710
Supplier:   Mopec
Supplier #:  AB044
Forceps, Dressing, Mortech
Catalog # 76548-710
Supplier:  Mopec
Forceps, Dressing, Mortech
Forceps, Dressing, Mortech
6"
Price
$28.34
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dressing forceps are ideal for all post-mortem examinations and dissection procedures.
  • Standard grade stainless steel
  • Ribbed grip
  • Smooth tip
Specifications
  • Length:
    6"