Orders LinkContactUsLinkComponent
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
4"
Catalog # 76548-696
Supplier:   Mopec
Supplier #:  AB037
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
Catalog # 76548-696
Supplier:  Mopec
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
Forceps, Iris Tissue, Straight 1×2 Teeth, Mortech
4"
Price
$22.02
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sharp 1×2 interlocking teeth for secure grip with minimal tissue compression.
  • Straight tips with 1×2 teeth
  • Standard grade stainless steel
Specifications
  • Length:
    4"