ContactUsLinkComponent
Dissecting Kit, Anatomy, Mortech
Dissecting Kit, Anatomy, Mortech
  76548-510
undefined
Dissecting Kit, Anatomy, Mortech
  76548-510
 :  AA305
  • Cat. No.:
    76548-510
Includes 10 standard instruments in a vinyl case with elasticized straps to perform a variety of post-mortem dissection needs.
 : Kit contains one of each instrument listed: model AA008 dissecting scissors, straight, 5.5"; BA021 Mayo scissors, curved, 5.5", AB003 dressing forceps, 5.5", AB011 tissue forceps, 1×2 Teeth, 5.5", AC003 Kelley forceps, curved, 5.5", AD004 grooved director, plain end, 5.5", AD029 triple chain hook, AD041 Mayo-hegar needle holder, 6", AJ101 knife handle #4.