Human Recombinant CD19 (from HEK293), PE Labeled Protein
Catalog # 103871-440
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:PE (Phycoerythrin)
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:200 tests
- Tag sequence:His
- Storage Conditions:–20 °C, avoid light
- Biological Activity:1e6 of the anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human CD19 (20-291), His Tag (Cat. No. CD9-HP2H3). PE Streptavidin was used as negative control (QC tested).
- Gene ID:P15391-1;930;NP_001761.3
- Amino Acid Number:Pro 20 - Lys 291
- Protein Synonyms:CD19,B4,CVID3,MGC12802
- Protein/Peptide Name:CD19
- Molecular Weight:33.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103871-440
Specifications
About this item
PE-Labeled Human CD19 (20-291), His Tag (CD9-HP2H3) is produced via site-specific conjugation of PE to Human CD19 (20-291), His Tag under optimal conditions with a proprietary technology. Human CD19 (20-291), His Tag is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291.
- Predicted N-terminus: Pro 20