Hexagonal Mass Set of 5, Plated
470339-280
- Description:Hexagonal mass set of 5, plated
- Cat. No.:470339-280
Zinc plated hexagonal masses.
- Set of five zinc-plated hexagonal masses
- Accurate and non-rolling
- Packaged in a sturdy cardboard box
- Suitable for ages 11+
Set of five zinc-plated hexagonal masses supplied in a sturdy cardboard box. Includes one each of 100 g, 500 g, and 1000 g, and two each of 200 g masses.