Deluxe Slotted Weight Set of 8, with Rack
Catalog # 470339-624
Supplier: United Scientific Supplies
Specifications
- Description:Deluxe slotted weight set of 8, with rack
- Cat. No.:470339-624
Specifications
About this item
Slotted brass weights for use with hangers.
- Set of 8 slotted brass weights
- Durable rack also included
- Also available as a set of 10 (WHST10) or 12 (WHST12)
Set of 8 high-quality slotted brass weights with durable rack.
Weights included: 1×500 g; 2× 200 g; 1×100 g; 1×50 g; 2×20 g; 1×10 g