Hooked Weight Set, Black Enamel
470339-458
- Description:Hooked weight set, black enamel
- Cat. No.:470339-458
Enameled hooked weights with a plastic storage case.
- Set of nine hooked cast iron weights with black enamel finish
- Weights can be hooked in tandem
- Sturdy plastic storage box included
Set of nine hooked cast iron masses with black enamel finish, shipped in a plastic storage base. Includes one each 10, 50, 100, 500, 1000 g, and two each 20 g and 200 g weights. Weights can be hooked in tandem.