Basic Spectroscope
Basic spectroscope
470338-940
: SPSCP1
Hand-held spectroscope shows the component wavelengths of light.
- Hand-held spectroscope
- Durable construction
An excellent hand-held spectroscope for viewing the spectra of many different light sources. Durable construction with compound prisms and converging lenses made of high-quality optical glass.
- Description:Basic spectroscope