ContactUsLinkComponent
Equal Mass Density Cylinders, Diverse Materials
Equal Mass Density Cylinders, Diverse Materials
Catalog # 470339-260
undefined
Equal Mass Density Cylinders, Diverse Materials
Catalog # 470339-260
Supplier Number:  SGMD05
Specifications
  • Description:
    Equal mass density cylinders, diverse materials, set of 5
  • Environmentally Preferable:
  • Cat. No.:
    470339-260
Specifications
About this item
Equal mass cylinders of metals and nonmetals for density investigations.
  • Set of 5 cylinders made of diverse materials, each with identical mass and radius
  • Ideal for lessons around specific gravity
  • Includes one each of acrylic, PVC, nylon, aluminum, and brass
  • Storage box and activity guide also included
Each specimen in this set of five diverse materials cylinders has identical mass and radius. Materials included are acrylic, PVC, nylon, aluminum, and brass. Includes a storage box and activity guide.