Density Blocks, Hooked
Catalog # 470339-884
Supplier: United Scientific Supplies
Specifications
- Description:Hooked density blocks
- Cat. No.:470339-884
Specifications
About this item
Large metal block with hooks for density or specific heat experiments.
- Set of 4 oversized blocks, ideal for demonstrating the density of different materials
- Includes one each of aluminum, brass, steel, and copper
- Each cube is 3.2 cm and features an attached hook
- Includes a reference table showing density values of individual specimens
The hooks embedded into these block allow you to easily submerge these into water for water displacement experiments. If you need to investigate specific heat, these metal block can be easily added into a calorimeter.