Secchi Disk
Secchi disk
Catalog # 470339-728
Supplier: United Scientific Supplies
Supplier #: SCDSK1
Jump to:
Product Details & Documents
See the clarity or turbidity of a water source.
- Ideal for water clarity measurements
- Made from durable white plastic, with alternating screen-printed black quadrants
- Includes attached sinker and rot-proof polymer line on polymer foam bobbin
Good-quality and economical Secchi disk for water clarity measurements. Constructed of robust white plastic disk with alternating screen-printed black quadrants. Includes attached sinker and rot-proof polymer line on polymer foam bobbin. Line length is approximately 72 feet (22 m), and includes a marking at each foot.
Specifications
- Description:Secchi disk