Pulley Demonstration Set
Catalog # 470339-092
Supplier: United Scientific Supplies
Specifications
- Description:Pulley demonstration set
- Cat. No.:470339-092
Specifications
About this item
Large demonstration kit for pulley systems, with wheel and axle.
- Convenient set for a variety of pulley demonstrations
- Also includes a 20 cm crossbar with clamp
- Accessories include multiple pulleys, clamps, and hooked weights
- Activity guide included
The durable 20×15 cm base is fitted with a capstan and a 61 cm tall aluminum support rod. Also includes a 20 cm crossbar with clamp. Includes activity guide.
Demonstration set includes: Wooden base with capstan, hook, and rod support; Vertical support rod (24"); Horizontal support rod with clamp (8"); 1 wheel and axle; 8 single sheave pulleys; 2 double sheave pulleys; 1 tommy bar; 1 clamp holder; 6 hook collar clamps; 2 pulley cords on spool; 1 hooked weight (1 kg); 1 hooked weight (500 g); 1 hooked weight (1 kg); 1 hooked weight (500 g); 2 hooked weights (200 g); 1 hooked weight (100 g); 1 hooked weight (50 g); 1 hooked weight (20 g); 1 hooked weight (10 g); Activity guide.