Orders LinkContactUsLinkComponent
Hardware Assortment
Hardware Assortment
Hardware assortment
Catalog # 470339-562
Supplier #:  HRDKIT1
Hardware Assortment
Catalog # 470339-562
Hardware Assortment
Hardware Assortment
Hardware assortment
Price
$149.60
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Quality assortment of hardware for 2 to 4 students.
  • See product description for full listing of contents
  • Shipped in a re-usable storage box
32-piece kit includes a variety of commonly used hardware items in a general science, physical science, or chemistry laboratory. Designed for use by 2 to 4 students. Shipped in a re-usable storage box.
Kit contents: 2 alligator clips with 24" wire; 1 burette brush; 1 burette clamp, double; 1 clamp boss head; 1 clay pipe triangle (2"); 1 crucible tong, plated (9"); 2 electrodes, carbon rods; 1 metric ruler, plastic (30 cm); 1 nichrome wire with aluminum handle; 1 glass-marking pencil (blue); 1 pinch clamp Mohr's (2.25"); 1 ring stand (5×8" base); 1 ring support with 4" clamp; 2 rubber stoppers, # 0, one-hole; 2 rubber stoppers, # 3, one-hole; 2 rubber stoppers, # 6, one-hole; 2 rubber stoppers, # 6, two-hole; Rubber tubing, (4'L, 0.25" I.D.); 2 test tube clamps; 1 test tube brush (3.5"); 1 wooden test tube rack (12 holes, 12 pins); 1 thermometer (20 to 150 °C); 1 polyethylene wash bottle (250 ml); 1 water reservoir, pneumatic trough; 1 wire gauze, ceramic (5×5").
Specifications
  • Description:
    Hardware assortment