ContactUsLinkComponent
Hooke's Law Apparatus
Hooke's Law Apparatus
Catalog # 470340-274
undefined
Hooke's Law Apparatus
Catalog # 470340-274
Supplier Number:  HKLW12
Specifications
  • Description:
    Hooke's law apparatus
  • Environmentally Preferable:
  • Cat. No.:
    470340-274
Specifications
About this item
Demonstrate the relationship between force and elongation in a spring.
  • Requires slotted weights
  • Includes activity guide
Quantitatively show the principles related to Hooke's Law with this complete apparatus. Includes a 14" metal rod that mounts on a 4.5" diameter base. Scale in centimeters and inches can be easily adjusted along the rod. The plated steel weight holder and spring are attached to an adjustable collar with crossbar. Requires slotted weights (not included - United Scientific WHST08 set recommended).