United Scientific Student Force Table
Catalog # 470339-000
Supplier: United Scientific Supplies
Specifications
- Description:Student force table
- Environmentally Preferable:
- Cat. No.:470339-000
Specifications
About this item
Illustrate force vectors and resultants.
- Complete student force table set features a dual 0.5 degree scale
- Lightweight, durable table top, with all-metal tripod stand
- Table diameter is 40.6 cm (16")
- Includes activity guide
This student force table is a complete set that features a dual scale; an outer scale for accurate pulley positioning, and an inner scale for orienting a tracing paper overlay. Set includes four complete weight sets (each with brass slotted weights, 2×5 g, 4×10 g, 4×20 g, and 20 g brass weight hanger), four pulleys with clamps and two sets of strings that allow students to investigate the equilibrium of either three or four forces. A 16" diameter lightweight, durable table top and sturdy cast iron support allow both ease of use and accuracy. Includes activity guide.