Filtering Kit
Catalog # 470339-072
Supplier: United Scientific Supplies
Specifications
- Description:Filtering kit
- Environmentally Preferable:
- Cat. No.:470339-072
Specifications
About this item
Compact kit for vacuum filtration.
- Complete kit, including hand vacuum pump, enables faster liquid filtration
- Hand vacuum pump with gauge also available separately (HVP001)
This easy-to-use set serves as a faster alternative to gravy-based liquid filtration.
Kit contents: Hand vacuum pump with gauge (HVP001), 500 ml filtering flask, 90 mm porcelain Buchner funnel, Pack of 100 filter papers (90 mm diameter), Rubber stopper, Vacuum hose, and Activity guide.