Free Fall Apparatus
Catalog # 470339-676
Supplier: United Scientific Supplies
Specifications
- Description:Free fall apparatus
- Environmentally Preferable:
- Cat. No.:470339-676
Specifications
About this item
Accurately measure the acceleration due to gravity through a 150 cm fall.
- Free fall apparatus for measuring the acceleration of gravity
- 150 cm fall length
- Requires digital timer DTPHG1 (not included)
This convenient free fall apparatus includes an electromagnetic ball release, a pair of photogates on a long cable harness, two steel balls, a plumb line, and a catcher. Requires a digital timer DTPHG1 (not included). Includes activity guide.