Order Entry
United States
ContactUsLinkComponent
Large U-Shaped Electromagnet
Large U-Shaped Electromagnet
Catalog # 470339-632
undefined
Large U-Shaped Electromagnet
Catalog # 470339-632
Supplier Number:  EMGT01

Specifications

  • Description:
    Large u-shaped electromagnet
  • Cat. No.:
    470339-632

Specifications

About this item

Demonstrate the power of an electromagnet.

  • Classroom electromagnet lifts up to 3.5 kg
  • Operates on 1.5 to 6 V DC

Operating on 1.5 to 6 V DC, this classroom electromagnet lifts up to 3.5 kg. A current direction indicator is included for the study of the magnetic field.