ContactUsLinkComponent
Electroscopes Kit
Electroscopes Kit
Catalog # 470339-244
undefined
Electroscopes Kit
Catalog # 470339-244
Supplier Number:  ELKIT1
Specifications
  • Description:
    Electroscopes kit
  • Cat. No.:
    470339-244
Specifications
About this item
Demonstrate electrostatic principles with these easy to build electroscopes.
  • Convenient kit for classroom demonstrations around key electrostatics concepts
  • Includes everything necessary to construct two flask form electroscopes
Demonstrate key electrostatics concepts with this kit. Contains everything necessary to construct two flask form electroscopes. Detailed manual included experiments using two electroscopes for positive and negative charge studies, and a Faraday's Pail experiment.
Kit contents: 2 glass flasks, 2 aluminum disc terminals, 2 sets of 4 foil leaves, 2 electrode assemblies, 1 aluminum can (Faraday pail), 1 hard rubber rod, 1 silk cloth, 1 acrylic rod, and 1 wool cloth.