Demo Diffraction Grating
Demo diffraction grating
Catalog # 470339-988
Supplier: United Scientific Supplies
Supplier #: DFG003
Jump to:
Product Details & Documents
Convenient diffraction grating with three different spacings
- Demonstration slide features three 20×10 mm gratings with 100, 300 and 600 lines per mm
- Mounted between glass plates
- Viewing area of each section is 15×10 mm
Use this diffraction grating to demonstrate the optical properties of three different diffraction gratings.
Specifications
- Description:Demo diffraction grating