Ballistics Car
470339-094
- Description:Ballistics car
- Cat. No.:470339-094
Demonstrate how an object in motion remains in motion.
- Easy-to-use tool to demonstrate the basics of ballistics
- Supplied with a steel ball, release pin, and cord
- Barrel permits two different heights of ball flight
- Activity guide included
Push the cart to eject the steel ball from the spring-loaded barrel. The ball falls back into the barrel of the moving car, demonstrating that the forward motion of the ball is the same as the vehicle from which it was ejected. Comes with a steel ball, release pin, and cord. The barrel permits two different heights of ball flight. Includes activity guide.