Wash Bottles, Unitary
Catalog # 470340-072
Supplier: United Scientific Supplies
Specifications
- Capacity:500 ml
- Description:Wash bottles, unitary
- Cat. No.:470340-072
Specifications
About this item
A highly visible red wash bottle.
- Colored red to provide a visual indicator of hazardous contents
- Made of translucent, pliable LDPE
- Dispense solutions easily with a light squeeze
- Graduated on outer surface at 100 ml intervals
Pack of four wash bottles with 500 ml capacity and built-in delivery tube. Red color provides a visual indicator that the bottle might contain hazardous liquids. Made of translucent, pliable low-density polyethylene (LDPE). Solutions can be easily dispensed with a light squeeze. Bottles are graduated on the outer surface at 100 ml intervals.