ContactUsLinkComponent
Utility Tray, Polypropylene
Utility Tray, Polypropylene
Catalog # 470340-028
undefined
Utility Tray, Polypropylene
Catalog # 470340-028
Supplier Number:  81721
Specifications
  • Description:
    Utility tray, polypropylene
  • Cat. No.:
    470340-028
Specifications
About this item
A deep polypropylene tray designed for all labware needs.
  • Autoclavable
  • Deep 5" tray
  • Tray dimensions: 15×14×5"
Steam autoclavable, polypropylene molded tray can be used for sterilizing and drying labware. Trays are also ideal for labware storage.