Order Entry
United States
ContactUsLinkComponent
 
DSP (Dithiobis(succinimidyl propionate)), No-Weigh™ Format, Pierce™
Catalog # 103523-924
Supplier:  Invitrogen
CAS Number:  
DSP (Dithiobis(succinimidyl propionate)), No-Weigh™ Format, Pierce™
Catalog # 103523-924
Supplier:  Invitrogen
Supplier Number:  A35393
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Size:
    10 x 1 mg
  • Cat. No.:
    103523-924

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    NHS Ester
  • Spacer:
    Cleavable, Long
  • Form:
    Powder
  • Quantity:
    50 mg / 1 g
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :404.41 g/mol
Storage Temperature:Refrigerator
CAS Number:57757-57-0
Formula:C₁₄H₁₆N₂O₈S₂
MDL Number:MFCD00042045