Sulfo-GMBS (N-Maleimidobutyryloxysulfosuccinimide ester)
Catalog # 82022-796
Supplier: G-Biosciences
CAS Number:
Specifications
- Size:100 mg
- Cat. No.:82022-796
- Supplier no.:BC14
Specifications
Specification Test Results
- Spacer Arm (Å):6.8
- Reactive Toward:Primary Amine + Sulfhydryl
- Membrane Permeable:No
- Water Soluble:Yes
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:360.297 g/mol
CAS Number:158018-86-1
Formula:C₁₂H₁₂N₂O₉S
MDL Number:MFCD00153605