Methyl hexanoate 99%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:106-70-7
:
Formula:C₇H₁₄O₂
MDL Number:MFCD00009510
:130.19 g/mol
Boiling Pt:151 °C (760 mmHg)
Melting Pt:–71 °C
Density:0.884 g/cm³
Flash Pt:42 °C
Storage Temperature:Ambient
UN:3272
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20 °C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together