Test Tubes, Plastic
plastic, Test tube, 12×75 mm, without cap; without graduation, round bottom
Catalog # 470332-708
Supplier: United Scientific Supplies
Supplier #: UNPLTUBE1275-PK/400
Jump to:
Product Details & Documents
Economical plastic test tubes.
- Rimless
- Rounded bottom
This 12×75 mm plastic test tube has a capacity of 5 ml. The tube features a round bottom.
Specifications
- Description:Test tube, 12×75 mm
- Material:plastic
- Tube Bottom Shape:round bottom
- Style:without cap; without graduation
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



