Soft Care® Sentry Handwash
Foaming antibacterial skin cleanser, refill, 6x 1300 ml
Catalog # 76547-802
Supplier: DIVERSEY INC.
Supplier #: 101100320
Jump to:
Product Details & Documents
Soft Care® Sentry is a fast-acting, foaming antimicrobial hand wash with superior mildness.
- Quick and effective antibacterial formula helps reduce cross-contamination
- NSF E2 registered for use in food handling establishments
- Rubs easily onto hands and leaves no sticky residue
- Mild on skin, avoiding excessive cracking and dryness for even the most frequent users
- Dye-Free and Fragrance-Free
- Luxurious foaming application
The unique formula gently cleanses the skin while killing germs that cause infection.
Formulated with Benzalkonium Chloride, Soft Care® Sentry removes soil from hands without the loss of antimicrobial performance. Rinses quickly, leaving no residue on skin.
Soft Care® Sentry is the perfect hand wash for food handling establishments and healthcare facilities.
Specifications
- Capacity:6x 1300 ml
- Description:Foaming antibacterial skin cleanser, refill