ContactUsLinkComponent
Crew Non-Acid Bowl & Bathroom Disinfectant Cleaner
Crew Non-Acid Bowl & Bathroom Disinfectant Cleaner
Catalog # 76547-796
Supplier:  DIVERSEY INC.
undefined
Crew Non-Acid Bowl & Bathroom Disinfectant Cleaner
Catalog # 76547-796
Supplier:  DIVERSEY INC.
Supplier Number:  100925283
Specifications
  • Size:
    12 x 32 oz
  • Cat. No.:
    76547-796
  • Packaging:
    Plastic bottle
Specifications
About this item
Crew® Non-Acid Disinfectant Cleaner is a neutral non-acid, multi-surface restroom disinfectant cleaner.
  • Non-abrasive ready-to-use liquid disinfectant with a fresh scent
  • Kills HIV, HCV, and HBV on pre-cleaned hard non-porous surfaces/objects previously soiled with blood and body fluids
  • Product eliminates odors and quickly cleans by removing dirt, grime, mold, mildew, body oils, dead skin, and blood
  • Meet OSHA's bloodborne pathogen standard and kills bacteria and viruses in 10 minutes
  • Great for daily cleaning with a simple ready to use flip top bottle
Cleans, disinfects and deodorizes toilet bowls, urinals, sinks, faucets, countertops and other hard, nonporous bathroom surfaces in one easy step.
Blue in color with a fresh scent.