- Environmentally Preferable:
- Cat. No.:76547-774
- Packaging:Plastic can
- No. of towels/wipes per container:4×160/pack
Avert® Sporicidal Disinfectant Cleaner Wipes cleans, disinfects and deodorizes hard, nonporous surfaces in one labor saving step with no rinsing required.
- Effective in 4 minutes against Clostridium difficile (C. diff) spores. Cleans, disinfects, and deodorizes in one labor-saving step
- Bleach-based formula means no quat-binding concerns. 1 minute contact time effective against a broad spectrum of pathogens including bacteria, antibiotic resistant bacteria, viruses, and fungi
- Meets minimum active ingredient requirements for CDC, SHEA, APIC and AORN
Designed for use on hard, nonporous surfaces such as those found in hospitals and other healthcare facilities.
Effective in 4 minutes against Clostridium difficile (C. diff) spores.
May be used to clean and disinfect sinks, faucets, fixtures, bathrooms, restrooms, tubs, and showers.