ContactUsLinkComponent
Avert® Sporicidal Disinfectant Cleaner
Avert® Sporicidal Disinfectant Cleaner
Catalog # 76547-768
Supplier:  DIVERSEY INC.
undefined
Avert® Sporicidal Disinfectant Cleaner
Catalog # 76547-768
Supplier:  DIVERSEY INC.
Supplier Number:  100842725
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    12 x 32 oz
  • Environmentally Preferable:
  • Cat. No.:
    76547-768
  • Packaging:
    Plastic bottle
Specifications
About this item
Avert® Sporicidal Disinfectant Cleaner cleans, disinfects and deodorizes hard, nonporous surfaces in one labor saving step with no rinsing required.
  • Effective in 4 minutes against Clostridium difficile (C. diff) spores. Cleans, disinfects, and deodorizes in one labor-saving step
  • Bleach-based formula means no quat-binding concerns. 1 minute contact time effective against a broad spectrum of pathogens including bacteria, antibiotic resistant bacteria, viruses, and fungi
  • Meets minimum active ingredient requirements for CDC, SHEA, APIC and AORN
Designed for use on hard, nonporous surfaces such as those found in hospitals and other healthcare facilities.
Effective in 4 minutes against Clostridium difficile (C. diff) spores.
May be used to clean and disinfect sinks, faucets, fixtures, bathrooms, restrooms, tubs, and showers.