Divoflow TC86, Low Foam Chlorinated Detergent for Hard Water
Catalog # 76547-754
Supplier: DIVERSEY INC.
Specifications
- Size:1 x 55 gal.
- Cat. No.:76547-754
- Packaging:Drum
Specifications
About this item
Divoflow TC 86 cleaner is a low foaming, chlorinated alkaline detergent for CIP and spray washing use in the food, beverage and dairy industries.
- Highly efficient in the removal of organic material including food soil, stains and off-flavors
- Low foaming under conditions of high turbulence
- Good hard water control and free rinsing, leaves no hard water films, minimizes acid cleaning cycles in most clean-in-place (CIP) operations
- Free-rinsing, minimizes water usage
- Liquid can be fed and controlled automatically in CIP systems