ContactUsLinkComponent
Acifoam VF10, Acidic Foam Cleaner-Descaler
Acifoam VF10, Acidic Foam Cleaner-Descaler
Catalog # 76547-752
Supplier:  DIVERSEY INC.
undefined
Acifoam VF10, Acidic Foam Cleaner-Descaler
Catalog # 76547-752
Supplier:  DIVERSEY INC.
Supplier Number:  57746100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    1 x 5 gal.
  • Environmentally Preferable:
  • Cat. No.:
    76547-752
  • Packaging:
    Pail
Specifications
About this item
Acifoam cleaner is based on inorganic acid and a blend of high foaming wetting agents/emulsifiers
  • Effective scale remover.
  • Versatile - Suitable for use on aluminum
  • Easy to use & Free-rinsing
  • Foam application extends contact time with vertical surfaces
It is specifically formulated for the removal of water hardness scale and other mineral deposits.
This product is also recommended for the periodic cleaning and descaling of high temperature processing equipment, such as cooking vessels, and also the exteriors of filler equipment, storage tanks and conveyers.
It is also effective for daily application to break down protein deposits in the fishing industry.
This product is suitable for use with a wide range of foam application equipment