Divosan® Spectrum Sanitizer / Disinfectant
Supplier: DIVERSEY INC.
Certificates
About this item
Divosan® Spectrum is a concentrated neutral liquid quaternary ammonium sanitizer providing effective bacteriological control in a wide variety of beverage, dairy and food processing operations.
- Liquid formulation: Mixes immediately and can be controlled automatically. Easily dispensed for accuracy and control. Heat stable: Suitable for warm and hot application where necessary. Non-corrosive: Can be used on all surfaces.
- Natural deodorizer: Leaves cleaned areas smelling fresh by eliminating odors.
- Broad spectrum kill: Blend of two quats more effectively controls bacteria, yeasts and molds.
- Residual properties: Produces bacteriostatic effect to retard bacterial growth.