Orders LinkContactUsLinkComponent
SCA Sentol®, Acid Cleaner
SCA Sentol®, Acid Cleaner
1 x 55 gal., Drum
Catalog # 76547-722
Supplier:   DIVERSEY INC.
Supplier #:  9676470
SCA Sentol®, Acid Cleaner
Catalog # 76547-722
Supplier:  DIVERSEY INC.
SCA Sentol®, Acid Cleaner
1 x 55 gal., Drum
Price
$1,950.36
Log in for availability
Carton of 55
Product Details & Documents
SCA Sentol® cleaner is a phosphoric acid-based low foaming liquid with excellent penetration and cleaning abilities for use on pharmaceutical and biotechnology processing equipment.
  • Penetrates and dissolves mineral deposits quickly and thoroughly
  • Optimum blend of cleaning/wetting agents provide rapid penetration and vigorous cleaning action
  • Free-rinsing for a streak-free and bright finish
  • Safe on stainless steel, glass and copper, when used as directed
  • Suitable for all application, methods, spray, soak circulation or manual
Documents
Specifications
  • Size:
    1 x 55 gal.
  • Packaging:
    Drum