Divosan® Forte, 30 gal
Divosan® Forte, 1 x 30 gal., Drum
Catalog # 76547-708
Supplier: DIVERSEY INC.
Supplier #: 08277070
Jump to:
Product Details & Documents
Divosan® Forte sanitizer is a non-foaming and 15% peracetic acid based liquid.
- Effective at lower temperatures, saving on energy costs
- Can be used under CO2 atmospheres
- Breakdown products are acetic acid and water
- Pungent odor alerts operator of any spills
- Can be used in most food/beverage systems when used as directed
- Non-foaming formula provides trouble-free spray, clean-in-place (CIP) and evaporator cleaning
It is used for sanitizing tanks, pipelines, evaporators, fillers, pasteurizers and aseptic equipment in dairies, wineries, breweries, beverage plants and for other nonporous food contact sanitizing applications.
Documents
Specifications
- Description:Divosan® Forte
- Size:1 x 30 gal.
- Packaging:Drum