ContactUsLinkComponent
Heavy Duty Liquid Alkaline Cleaner
Heavy Duty Liquid Alkaline Cleaner
Catalog # 76547-686
Supplier:  Avantor
Heavy Duty Liquid Alkaline Cleaner
Catalog # 76547-686
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    Bulk
  • Environmentally Preferable:
  • Cat. No.:
    76547-686
Specifications
About this item
Liquid BR-5512-LAB is a concentrated light yellow, heavy duty alkaline cleaning compound designed to remove carbon, oils, grease, and other tenacious soils.
  • Effective in removing carbon oils, greases, and proteinaceous deposits
  • It can be used in central cleaning systems, soaking operations, immersion tanks, spray washers, and re-circulating systems
Liquid BR-5512-LAB is a concentrated light yellow, heavy duty alkaline cleaning compound designed to remove carbon, oils, grease, and other tenacious soils. It is manufactured and packaged especially for the pharmaceutical industry.