
Sodium dehydracetate 99%
Catalog # TS36485-5000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:500 g
- Cat. No.:TS36485-5000
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Titration with HClO4:≥98.5 %
Specification Test Results
About this item
:
:190.13 g/mol
Melting Pt:295 °C
EINECS:224-580-1
:GHS07
Storage Temperature:Ambient
CAS Number:4418-26-2
Formula:C₈H₇NaO₄
MDL Number:MFCD00040583