Order Entry
United States
ContactUsLinkComponent
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Corrosive
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Catalog # TS46068-0010
CAS Number:  
undefined
Corrosive
Copper (II) ethylenediamine 1 M in water, Technical Grade not for viscosity determination
Catalog # TS46068-0010
Supplier Number:  46068-0010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    TS46068-0010

Specifications

Specification Test Results

  • Appearance:
    Dark blue Solution
  • Strength:
    0.98 to 1.02 M (copper molarity)

Specification Test Results

About this item

 :
 :217.76 g/mol
Density:1.1 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:1761
ADR:8,II
 :GHS05
CAS Number:14552-35-3
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number:MFCD00274628