Solvent Resistant 2-Stop Tubing
Supplier: VWR International
Certificates
About this item
Solvent Resistant, Flared Two-Stop PVC pump tubing.
Tubing for peristaltic pump application where solvent resistance is required.
Buy 5 packs and receive a 10% discount*
Buy 10 packs and receive a 20% discount*
Buy 20 packs and receive a 25% discount*
*Additional discounts do not apply
- Environmentally Preferable:
Frequently Bought Together