Silastic® Laboratory Transfer Pump Tubing
Supplier: Avantor
Certificates
About this item
Platinum-cured tubing for a wide range of applications.
- Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotech, filtration, and environmental analysis
Characteristics: Platinum-cured, nonpyrogenic, bacteria and fungus-resistant, extremely low extractables, and does not impart a taste or odor
Certifications: Exceeds USP Class VI Plastics Test requirements, elastomer and tubing are manufactured under Bulk Pharmaceutical cGMPs in a FDA compliant, ISO-9002-certified facility
Temperature range: –65 °F to 480 °F (–54 °C to 249 °C)
Sterilize: Autoclave or radiation.
- Environmentally Preferable:
Related Information
Frequently Bought Together