Parker Hannifin Flaring Tools for PFA Flare Fittings
Supplier: Avantor
Certificates
About this item
Chemically-inert PFA handles most aggressive chemicals.
- Optimize performance of your PFA Flare Fittings
Use these tools for a better fit. Heat the PFA tubing with a heat gun, and insert the tool to create the flare. Tools are available for any size of tubing from 1/4-inch to 1-inch outer diameter.
- Environmentally Preferable:
Frequently Bought Together